siRNA Details
| Gene Symbol: Cdh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053393.2 | NP_445845.2 | 2 | 547 | ||
| GenBank mRNA | AB010436.1 | - | 2 | 547 | ||
| AB010437.1 | - | 2 | 547 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdh8| Gene Symbol: Cdh8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053393.2 | NP_445845.2 | 2 | 547 | ||
| GenBank mRNA | AB010436.1 | - | 2 | 547 | ||
| AB010437.1 | - | 2 | 547 | |||