siRNA Details
| Gene Symbol: Gfra3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053398.1 | NP_445850.1 | 2 | 330 | ||
| GenBank mRNA | BC079378.1 | - | 2 | 330 | 7110848 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gfra3| Gene Symbol: Gfra3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053398.1 | NP_445850.1 | 2 | 330 | ||
| GenBank mRNA | BC079378.1 | - | 2 | 330 | 7110848 | |