siRNA Details
| Gene Symbol: Gfra3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053398.1 | NP_445850.1 | 3 | 461 | ||
| GenBank mRNA | BC079378.1 | - | 3 | 461 | 7110848 | |
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_010280, AF051766, Y15110, AF036163, AB008833, and others.

Gfra3
Genomic Map