siRNA Details
| Gene Symbol: Kcnk9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053405.2 | NP_445857.2 | 2 | 420 | ||
| GenBank mRNA | AF192366.1 | - | 2 | 510 | ||
| AF257082.1 | - | 2 | 425 | |||
| AF391084.1 | - | 2 | 420 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnk9| Gene Symbol: Kcnk9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053405.2 | NP_445857.2 | 2 | 420 | ||
| GenBank mRNA | AF192366.1 | - | 2 | 510 | ||
| AF257082.1 | - | 2 | 425 | |||
| AF391084.1 | - | 2 | 420 | |||