siRNA Details
| Gene Symbol: Pomt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053406.2 | NP_445858.1 | 7 | 729 | ||
| XM_006234001.2 | XP_006234063.1 | 7 | 1331 | |||
| XM_008761776.1 | XP_008759998.1 | 5 | 439 | |||
| GenBank mRNA | AF192388.1 | - | 7 | 702 | ||
| BC070912.1 | - | 7 | 699 | 7104668 | ||

Pomt1
Genomic Map