siRNA Details
| Gene Symbol: Chst3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053408.1 | NP_445860.1 | 2 | 169 | ||
| XM_006256427.2 | XP_006256489.1 | 2 | 283 | |||
| XM_006256428.2 | XP_006256490.1 | 3 | 315 | |||
| XM_008772929.1 | XP_008771151.1 | 3 | 820 | |||
| GenBank mRNA | AF178689.2 | - | 2 | 169 | ||

Chst3
Genomic Map