siRNA Details
Gene Symbol: Maged1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053409.2 | NP_445861.1 | 3 | 836 | ||
GenBank mRNA | AF217964.1 | - | 3 | 972 | ||
AF274043.1 | - | 1 | 175 | |||
AJ133038.1 | - | 1 | 299 | |||
BC081756.1 | - | 3 | 836 | 7134252 |
