siRNA Details
Gene Symbol: Snx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053411.1 | NP_445863.1 | 7, 8 | 804 | ||
XM_006243317.2 | XP_006243379.1 | 7, 8 | 687 | |||
XM_008766349.1 | XP_008764571.1 | 7, 8 | 859 | |||
GenBank mRNA | AF218916.1 | - | 7, 8 | 804 | ||
BC092201.1 | - | 7, 8 | 807 | 7309531 |
