siRNA Details
Gene Symbol: Ilf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053412.1 | NP_445864.1 | 18, 19 | 2282 | ||
XM_008765990.1 | XP_008764212.1 | 18, 19 | 2417 | |||
XM_008765991.1 | XP_008764213.1 | 18, 19 | 2415 | |||
XM_008765992.1 | XP_008764214.1 | 18, 19 | 2402 | |||
XM_008765993.1 | XP_008764215.1 | 17, 18 | 2378 | |||
XM_008765994.1 | XP_008764216.1 | 17, 18 | 2363 | |||
GenBank mRNA | AF220102.1 | - | 18, 19 | 2282 |
