siRNA Details
Gene Symbol: Ilf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053412.1 | NP_445864.1 | 15 | 1841 | ||
XM_008765990.1 | XP_008764212.1 | 15 | 1976 | |||
XM_008765991.1 | XP_008764213.1 | 15 | 1974 | |||
XM_008765992.1 | XP_008764214.1 | 15 | 1961 | |||
XM_008765993.1 | XP_008764215.1 | 14 | 1937 | |||
XM_008765994.1 | XP_008764216.1 | 14 | 1922 | |||
XM_008765995.1 | XP_008764217.1 | 15 | 1976 | |||
XM_008765996.1 | XP_008764218.1 | 15 | 1961 | |||
GenBank mRNA | AF220102.1 | - | 15 | 1841 |
