siRNA Details
| Gene Symbol: Strbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053416.2 | NP_445868.2 | 17 | 2146 | ||
| XM_006234108.2 | XP_006234170.2 | 18 | 2484 | |||
| XM_008761777.1 | XP_008759999.1 | 18 | 2184 | |||
| GenBank mRNA | BC078774.1 | - | 16 | 1768 | ||
| BU759091.1 | - | 1 | 426 | |||
| DY320620.1 | - | 5 | 460 | |||
| FQ232581.1 | - | 7 | 815 | |||

Strbp
Genomic Map