siRNA Details
| Gene Symbol: Strbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053416.2 | NP_445868.2 | 9 | 1137 | ||
| XM_006234108.2 | XP_006234170.2 | 10 | 1475 | |||
| XM_008761777.1 | XP_008759999.1 | 10 | 1175 | |||
| XM_008761778.1 | XP_008760000.1 | 10 | 1175 | |||
| GenBank mRNA | BC078774.1 | - | 8 | 759 | ||
| CA512540.1 | - | 6 | 566 | |||

Strbp
Genomic Map