siRNA Details
| Gene Symbol: Foxo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191846.2 | NP_001178775.1 | 2 | 1401 | ||
| GenBank mRNA | AF247812.1 | - | 2 | 792 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Foxo1| Gene Symbol: Foxo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191846.2 | NP_001178775.1 | 2 | 1401 | ||
| GenBank mRNA | AF247812.1 | - | 2 | 792 | ||