siRNA Details
| Gene Symbol: Sf3b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053426.1 | NP_445878.1 | 20 | 3125 | ||
| XM_006244933.2 | XP_006244995.1 | 20 | 3122 | |||
| XM_006244934.2 | XP_006244996.1 | 17 | 4030 | |||
| GenBank mRNA | FQ234614.1 | - | 2 | 178 | ||

Sf3b1
Genomic Map