siRNA Details
| Gene Symbol: Sf3b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053426.1 | NP_445878.1 | 14 | 2046 | ||
| XM_006244933.2 | XP_006244995.1 | 14 | 2043 | |||
| XM_006244934.2 | XP_006244996.1 | 11 | 2951 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sf3b1| Gene Symbol: Sf3b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053426.1 | NP_445878.1 | 14 | 2046 | ||
| XM_006244933.2 | XP_006244995.1 | 14 | 2043 | |||
| XM_006244934.2 | XP_006244996.1 | 11 | 2951 | |||