siRNA Details
| Gene Symbol: Fgf13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053428.1 | NP_445880.1 | 5 | 596 | ||
| XM_006257592.1 | XP_006257654.1 | 5 | 864 | |||
| XM_008773572.1 | XP_008771794.1 | 6 | 772 | |||
| XM_008773573.1 | XP_008771795.1 | 6 | 834 | |||
| GenBank mRNA | AF271786.1 | - | 5 | 596 | ||

Fgf13
Genomic Map