siRNA Details
| Gene Symbol: Fgf13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053428.1 | NP_445880.1 | 1 | 138 | ||
| GenBank mRNA | AF271786.1 | - | 1 | 138 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fgf13| Gene Symbol: Fgf13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053428.1 | NP_445880.1 | 1 | 138 | ||
| GenBank mRNA | AF271786.1 | - | 1 | 138 | ||