siRNA Details
| Gene Symbol: Qsox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109898.1 | NP_001103368.1 | 9 | 1090 | ||
| NM_053431.3 | NP_445883.1 | 9 | 1090 | |||
| GenBank mRNA | AB044285.1 | - | 9 | 1090 | ||
| AF285078.1 | - | 9 | 1112 | |||
| AY623665.1 | - | 9 | 1092 | |||

Qsox1
Genomic Map