siRNA Details
| Gene Symbol: Stau1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109907.1 | NP_001103377.1 | 5 | 613 | ||
| NM_053436.2 | NP_445888.1 | 5 | 631 | |||
| GenBank mRNA | AF227200.1 | - | 5 | 618 | ||
| AF290989.1 | - | 5 | 600 | |||
| AJ010200.1 | - | 3 | 294 | |||
| BC081755.1 | - | 5 | 571 | |||
| BC101858.1 | - | 5 | 609 | 7937343 | ||
| DN932990.1 | - | 5 | 642 | |||

Stau1
Genomic Map