siRNA Details
| Gene Symbol: Dgat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053437.1 | NP_445889.1 | 7 | 692 | ||
| GenBank mRNA | AF296131.1 | - | 7 | 692 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dgat1| Gene Symbol: Dgat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053437.1 | NP_445889.1 | 7 | 692 | ||
| GenBank mRNA | AF296131.1 | - | 7 | 692 | ||