siRNA Details
Gene Symbol: Slco1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053441.1 | NP_445893.1 | 11 | 1894 | ||
XM_006237602.1 | XP_006237664.1 | 12 | 2042 | |||
GenBank mRNA | AF306546.1 | - | 11 | 1894 |
