siRNA Details
Gene Symbol: Slc7a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053442.1 | NP_445894.1 | 8 | 1668 | ||
GenBank mRNA | AB024400.1 | - | 8 | 1668 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc7a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053442.1 | NP_445894.1 | 8 | 1668 | ||
GenBank mRNA | AB024400.1 | - | 8 | 1668 |