siRNA Details
| Gene Symbol: Slc7a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053442.1 | NP_445894.1 | 10 | 2031 | ||
| GenBank mRNA | AB024400.1 | - | 10 | 2031 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc7a8| Gene Symbol: Slc7a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053442.1 | NP_445894.1 | 10 | 2031 | ||
| GenBank mRNA | AB024400.1 | - | 10 | 2031 | ||