siRNA Details
| Gene Symbol: Plcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053456.1 | NP_445908.1 | 6 | 3651 | ||
| XM_008767141.1 | XP_008765363.1 | 5 | 7432 | |||
| GenBank mRNA | D45920.1 | - | 6 | 3651 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plcl1| Gene Symbol: Plcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053456.1 | NP_445908.1 | 6 | 3651 | ||
| XM_008767141.1 | XP_008765363.1 | 5 | 7432 | |||
| GenBank mRNA | D45920.1 | - | 6 | 3651 | ||