siRNA Details
| Gene Symbol: Pick1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053460.1 | NP_445912.1 | 8 | 730 | ||
| GenBank mRNA | AF327562.1 | - | 8 | 730 | ||
| AF373289.1 | - | 8 | 652 | |||
| AF542094.1 | - | 8 | 700 | |||
| AJ240083.2 | - | 8 | 772 | |||
| BC072689.1 | - | 9 | 733 | 7105747 | ||

Pick1
Genomic Map