siRNA Details
| Gene Symbol: Nfs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053462.2 | NP_445914.2 | 292 | |||
| XM_006235365.2 | XP_006235427.1 | 294 | ||||
| GenBank mRNA | AF336041.1 | - | 253 | |||
| BC089205.1 | - | 258 | ||||
| BC104699.1 | - | 292 | 7189705 | |||
| Gene Symbol: LOC100911034 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749609.2 | XP_003749657.1 | 3 | 297 | ||
| XM_006235451.2 | XP_006235513.1 | 3 | 294 | |||

Nfs1
Genomic Map