siRNA Details
| Gene Symbol: Sema6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053471.2 | NP_445923.1 | 11 | 1470 | ||
| XM_006244348.2 | XP_006244410.1 | 12 | 1939 | |||
| XM_006244349.2 | XP_006244411.1 | 11 | 1292 | |||
| GenBank mRNA | AB000776.1 | - | 11 | 1470 | ||

Sema6b
Genomic Map