siRNA Details
| Gene Symbol: Ptp4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053475.1 | NP_445927.1 | 4 | 762 | ||
| GenBank mRNA | AJ007016.1 | - | 4 | 762 | ||
| BC060549.1 | - | 4 | 834 | 6891377 | ||
| FQ226595.1 | - | 1 | 54 | |||
| FQ230760.1 | - | 4 | 344 | |||
| FQ231532.1 | - | 4 | 468 | |||

Ptp4a2
Genomic Map