siRNA Details
| Gene Symbol: Pex11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053487.2 | NP_445939.1 | 3 | 672 | ||
| XM_008759560.1 | XP_008757782.1 | 3 | 671 | |||
| GenBank mRNA | AJ224120.1 | - | 3 | 704 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pex11a| Gene Symbol: Pex11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053487.2 | NP_445939.1 | 3 | 672 | ||
| XM_008759560.1 | XP_008757782.1 | 3 | 671 | |||
| GenBank mRNA | AJ224120.1 | - | 3 | 704 | ||