siRNA Details
| Gene Symbol: Xpo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053490.1 | NP_445942.1 | 20, 21 | 2821 | ||
| XM_006251578.2 | XP_006251640.1 | 21, 22 | 3617 | |||
| XM_008770437.1 | XP_008768659.1 | 19, 20 | 3145 | |||
| GenBank mRNA | AB105193.1 | - | 20, 21 | 2821 | ||
| AJ238278.1 | - | 9, 10 | 1483 | |||

Xpo1
Genomic Map