siRNA Details
| Gene Symbol: Plg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053491.2 | NP_445943.1 | 19 | 2442 | ||
| GenBank mRNA | AJ242649.1 | - | 19 | 2445 | ||
| BC091135.1 | - | 19 | 2442 | 7369357 | ||
| FQ210878.1 | - | 19 | 2585 | |||
| FQ218248.1 | - | 6 | 685 | |||

Plg
Genomic Map