siRNA Details
| Gene Symbol: Hacl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053493.1 | NP_445945.1 | 5, 6 | 493 | ||
| GenBank mRNA | AJ517469.1 | - | 5, 6 | 493 | ||
| BC078697.1 | - | 5, 6 | 477 | 7110355 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hacl1| Gene Symbol: Hacl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053493.1 | NP_445945.1 | 5, 6 | 493 | ||
| GenBank mRNA | AJ517469.1 | - | 5, 6 | 493 | ||
| BC078697.1 | - | 5, 6 | 477 | 7110355 | ||