siRNA Details
Gene Symbol: Entpd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053498.1 | NP_445950.1 | 2 | 207 | ||
XM_006235218.2 | XP_006235280.1 | 2 | 302 | |||
GenBank mRNA | AJ277748.1 | - | 2 | 207 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Entpd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053498.1 | NP_445950.1 | 2 | 207 | ||
XM_006235218.2 | XP_006235280.1 | 2 | 302 | |||
GenBank mRNA | AJ277748.1 | - | 2 | 207 |