siRNA Details
| Gene Symbol: Slc25a27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053500.1 | NP_445952.1 | 7 | 819 | ||
| XM_006244610.2 | XP_006244672.1 | 7 | 935 | |||
| XM_006244611.2 | XP_006244673.1 | 7 | 935 | |||
| XM_006244612.2 | XP_006244674.1 | 7 | 935 | |||
| XM_008766882.1 | XP_008765104.1 | 7 | 1155 | |||
| GenBank mRNA | AJ300162.1 | - | 7 | 819 | ||
| AJ300163.1 | - | 7 | 906 | |||
| AJ300164.1 | - | 7 | 927 | |||
| AY512661.1 | - | 7 | 749 | |||

Slc25a27
Genomic Map