siRNA Details
Gene Symbol: Slc25a27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053500.1 | NP_445952.1 | 9 | 1006 | ||
GenBank mRNA | AJ300162.1 | - | 9 | 1006 | ||
AY512661.1 | - | 9 | 936 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc25a27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053500.1 | NP_445952.1 | 9 | 1006 | ||
GenBank mRNA | AJ300162.1 | - | 9 | 1006 | ||
AY512661.1 | - | 9 | 936 |