siRNA Details
| Gene Symbol: Slc25a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053501.2 | NP_445953.1 | 2, 3 | 263 | ||
| GenBank mRNA | AF300424.1 | - | 2, 3 | 264 | ||
| AJ300165.1 | - | 1, 2 | 69 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc25a14| Gene Symbol: Slc25a14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053501.2 | NP_445953.1 | 2, 3 | 263 | ||
| GenBank mRNA | AF300424.1 | - | 2, 3 | 264 | ||
| AJ300165.1 | - | 1, 2 | 69 | |||