siRNA Details
| Gene Symbol: Slc24a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY009158.1 | - | 10 | 739 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc24a3| Gene Symbol: Slc24a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY009158.1 | - | 10 | 739 | ||