siRNA Details
| Gene Symbol: Folh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057185.2 | NP_476533.1 | 18 | 2094 | ||
| GenBank mRNA | AF039707.1 | - | 12 | 1180 | ||
| AF040256.1 | - | 18 | 2100 | |||
| AF513486.2 | - | 18 | 2094 | |||
| U75973.1 | - | 18 | 2051 | |||

Folh1
Genomic Map