siRNA Details
| Gene Symbol: Folh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_057185.2 | NP_476533.1 | 14, 15 | 1594 | ||
| GenBank mRNA | AF039707.1 | - | 8, 9 | 680 | ||
| AF040256.1 | - | 14, 15 | 1600 | |||
| AF513486.2 | - | 14, 15 | 1594 | |||
| U75973.1 | - | 14, 15 | 1551 | |||

Folh1
Genomic Map