siRNA Details
| Gene Symbol: Prkd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276715.1 | NP_001263644.1 | 13 | 1795 | ||
| XM_006240074.2 | XP_006240136.1 | 12 | 2053 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prkd1| Gene Symbol: Prkd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276715.1 | NP_001263644.1 | 13 | 1795 | ||
| XM_006240074.2 | XP_006240136.1 | 12 | 2053 | |||