siRNA Details
| Gene Symbol: Rhoq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053522.1 | NP_445974.1 | 3 | 610 | ||
| GenBank mRNA | AB031482.1 | - | 3 | 610 | ||
| BC061760.1 | - | 3 | 600 | 5621223 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rhoq| Gene Symbol: Rhoq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053522.1 | NP_445974.1 | 3 | 610 | ||
| GenBank mRNA | AB031482.1 | - | 3 | 610 | ||
| BC061760.1 | - | 3 | 600 | 5621223 | ||