siRNA Details
| Gene Symbol: Ddx52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053525.1 | NP_445977.1 | 10 | 1306 | ||
| GenBank mRNA | AB055628.1 | - | 10 | 1306 | ||
| BC089107.1 | - | 10 | 1308 | 7310368 | ||
| EF121975.1 | - | 10 | 1300 | |||
| EF121976.1 | - | 10 | 1300 | |||

Ddx52
Genomic Map