siRNA Details
| Gene Symbol: Smad9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138872.1 | NP_620227.1 | 980 | |||
| XM_008761009.1 | XP_008759231.1 | 3 | 1775 | |||
| GenBank mRNA | AF012347.1 | - | 980 | |||
| Gene Symbol: LOC103691556 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761019.1 | XP_008759241.1 | 1775 | |||
| XM_008761020.1 | XP_008759242.1 | 1775 | ||||

Smad9
Genomic Map