siRNA Details
| Gene Symbol: Smad9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138872.1 | NP_620227.1 | 351 | |||
| GenBank mRNA | AF012347.1 | - | 351 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smad9| Gene Symbol: Smad9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138872.1 | NP_620227.1 | 351 | |||
| GenBank mRNA | AF012347.1 | - | 351 | |||