siRNA Details
Gene Symbol: Smad9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138872.1 | NP_620227.1 | 1040 | |||
XM_008761009.1 | XP_008759231.1 | 3, 4 | 1835 | |||
GenBank mRNA | AF012347.1 | - | 1040 |
Gene Symbol: LOC103691556 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761019.1 | XP_008759241.1 | 1835 | |||
XM_008761020.1 | XP_008759242.1 | 1835 |
