siRNA Details
| Gene Symbol: Nbn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138873.2 | NP_620228.1 | 10 | 1329 | ||
| XM_006237916.1 | XP_006237978.1 | 10 | 1372 | |||
| GenBank mRNA | AF218575.1 | - | 10 | 1333 | ||
| BC085700.1 | - | 10 | 1297 | 7130978 | ||

Nbn
Genomic Map