siRNA Details
| Gene Symbol: Gna15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053542.1 | NP_445994.1 | 2 | 332 | ||
| XM_006241021.2 | XP_006241083.1 | 2 | 707 | |||
| GenBank mRNA | AB015308.1 | - | 2 | 407 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gna15| Gene Symbol: Gna15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053542.1 | NP_445994.1 | 2 | 332 | ||
| XM_006241021.2 | XP_006241083.1 | 2 | 707 | |||
| GenBank mRNA | AB015308.1 | - | 2 | 407 | ||