siRNA Details
| Gene Symbol: Lbr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134453.1 | NP_604448.1 | 8 | 1401 | ||
| GenBank mRNA | AB002466.1 | - | 8 | 1401 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lbr| Gene Symbol: Lbr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134453.1 | NP_604448.1 | 8 | 1401 | ||
| GenBank mRNA | AB002466.1 | - | 8 | 1401 | ||