siRNA Details
| Gene Symbol: Ncdn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053543.2 | NP_445995.2 | 3 | 1358 | ||
| XM_006238908.1 | XP_006238970.1 | 3 | 1067 | |||
| XM_006238909.1 | XP_006238971.1 | 3 | 1128 | |||
| GenBank mRNA | BC087000.1 | - | 3 | 1358 | 7190905 | |

Ncdn
Genomic Map