siRNA Details
| Gene Symbol: Sfrp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053544.1 | NP_445996.1 | 4 | 741 | ||
| GenBank mRNA | AF012891.1 | - | 4 | 741 | ||
| AF140346.1 | - | 4 | 892 | |||
| AF220608.1 | - | 989 | ||||
| FQ220237.1 | - | 1 | 50 | |||
| FQ229078.1 | - | 1 | 63 | |||
| FQ229224.1 | - | 3 | 172 | |||
More Information
Set Membership: |
|

Sfrp4
Genomic Map